Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tcaim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016864.3 | NP_001016864.1 | 9 - 10 | 1245 | 93 | |
XM_012964746.2 | XP_012820200.1 | 10 - 11 | 1383 | 93 | ||
XM_012964747.2 | XP_012820201.1 | 10 - 11 | 1412 | 93 | ||
XM_012964749.2 | XP_012820203.1 | 10 - 11 | 1415 | 93 | ||
XM_018094663.1 | XP_017950152.1 | 10 - 11 | 1316 | 93 | ||
GenBank mRNA | BC123041.1 | - | 8 - 9 | 1138 | 93 | |
CR848646.2 | - | 9 - 10 | 1247 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |