Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qtrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016891.2 | NP_001016891.1 | 2 - 3 | 344 | 76 | |
GenBank mRNA | CR848556.2 | - | 2 - 3 | 344 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |