Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept2-a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017039.2 | NP_001017039.1 | 8 - 9 | 785 | 87 | |
XM_012963217.2 | XP_012818671.1 | 9 - 10 | 996 | 87 | ||
GenBank mRNA | BC091009.1 | - | 8 - 9 | 785 | 87 | |
CR761939.2 | - | 8 - 9 | 750 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |