Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017116.2 | NP_001017116.1 | 6 - 7 | 931 | 102 | |
XM_012963315.2 | XP_012818769.1 | 5 - 6 | 1120 | 102 | ||
XM_012963318.2 | XP_012818772.1 | 6 - 7 | 926 | 102 | ||
XM_012963321.2 | XP_012818775.1 | 4 - 5 | 984 | 102 | ||
GenBank mRNA | BC168538.1 | - | 5 - 6 | 1010 | 102 | |
CR761183.2 | - | 6 - 7 | 931 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |