Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017205.2 | NP_001017205.1 | 7 - 8 | 1138 | 97 | |
GenBank mRNA | BC135154.1 | - | 7 - 8 | 1148 | 96 | |
CR760502.2 | - | 7 - 8 | 1138 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |