Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030330.1 | NP_001025501.1 | 10 - 11 | 1440 | 104 | |
GenBank mRNA | BC080175.1 | - | 10 | 1446 | 148 | |
CR760257.1 | - | 10 - 11 | 1440 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |