Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030330.1 | NP_001025501.1 | 7 - 8 | 1066 | 87 | |
GenBank mRNA | BC080175.1 | - | 7 - 8 | 1072 | 87 | |
BC161475.1 | - | 7 - 8 | 1059 | 87 | ||
CR760257.1 | - | 7 - 8 | 1066 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |