Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030421.2 | NP_001025592.2 | 5 - 6 | 1075 | 66 | |
XM_012961217.2 | XP_012816671.1 | 5 - 6 | 1198 | 66 | ||
GenBank mRNA | BC091041.1 | - | 5 - 6 | 975 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |