Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030472.1 | NP_001025643.1 | 12 - 13 | 1442 | 67 | |
XM_012963078.2 | XP_012818532.1 | 12 - 13 | 1568 | 67 | ||
XM_012963079.2 | XP_012818533.1 | 12 - 13 | 1541 | 67 | ||
XM_012963080.2 | XP_012818534.1 | 11 - 12 | 1512 | 67 | ||
GenBank mRNA | BC091612.1 | - | 12 - 13 | 1442 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |