Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030492.1 | NP_001025663.1 | 3 - 4 | 284 | 101 | |
GenBank mRNA | BC092545.1 | - | 3 - 4 | 284 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |