Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xkr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033943.1 | NP_001029115.1 | 1 - 2 | 700 | 93 | |
XM_018089355.1 | XP_017944844.1 | 3 - 4 | 1687 | 93 | ||
GenBank mRNA | AY699709.1 | - | 1 - 2 | 700 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |