Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011385.2 | NP_001011385.2 | 2 - 3 | 810 | 90 | |
XM_018092852.1 | XP_017948341.1 | 3 - 4 | 1190 | 90 | ||
XM_018092853.1 | XP_017948342.1 | 3 - 4 | 1088 | 90 | ||
XM_018092854.1 | XP_017948343.1 | 3 - 4 | 1140 | 90 | ||
XM_018092855.1 | XP_017948344.1 | 2 - 3 | 753 | 90 | ||
GenBank mRNA | CR855420.2 | - | 2 - 3 | 810 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |