Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015960.2 | NP_001015960.1 | 4 - 5 | 1267 | 95 | |
GenBank mRNA | BC161035.1 | - | 4 - 5 | 1383 | 95 | |
BC170673.1 | - | 4 - 5 | 1227 | 95 | ||
BC170677.1 | - | 4 - 5 | 1227 | 95 | ||
CR848438.2 | - | 4 - 5 | 1267 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |