Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016115.2 | NP_001016115.1 | 7 - 8 | 883 | 72 | |
GenBank mRNA | BC161036.1 | - | 7 - 8 | 879 | 72 | |
CR762101.2 | - | 7 - 8 | 883 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |