Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016403.2 | NP_001016403.1 | 6 - 7 | 579 | 67 | |
XM_012953064.1 | XP_012808518.1 | 6 - 7 | 967 | 67 | ||
XM_012953065.1 | XP_012808519.1 | 7 - 8 | 826 | 67 | ||
XM_012953066.2 | XP_012808520.1 | 7 - 8 | 805 | 67 | ||
XM_012953067.1 | XP_012808521.1 | 7 - 8 | 761 | 67 | ||
XM_012953069.2 | XP_012808523.1 | 7 - 8 | 740 | 67 | ||
XM_018092085.1 | XP_017947574.1 | 6 - 7 | 621 | 67 | ||
GenBank mRNA | BC170934.1 | - | 6 - 7 | 577 | 67 | |
BC170936.1 | - | 6 - 7 | 577 | 67 | ||
CR761141.2 | - | 6 - 7 | 579 | 67 | ||
FN562397.1 | - | 6 - 7 | 649 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |