Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gucd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016459.2 | NP_001016459.1 | 4 - 5 | 472 | 63 | |
GenBank mRNA | BC170918.1 | - | 4 - 5 | 463 | 63 | |
BC170920.1 | - | 4 - 5 | 463 | 63 | ||
CR760957.2 | - | 4 - 5 | 472 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |