Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016613.2 | NP_001016613.1 | 6 - 7 | 426 | 72 | |
XM_012959033.2 | XP_012814487.1 | 6 - 7 | 464 | 72 | ||
XM_018092088.1 | XP_017947577.1 | 7 - 8 | 512 | 72 | ||
GenBank mRNA | BC170594.1 | - | 5 - 6 | 416 | 72 | |
BC170598.1 | - | 5 - 6 | 416 | 72 | ||
CR760517.2 | - | 6 - 7 | 426 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |