Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ostc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016656.2 | NP_001016656.1 | 2 - 3 | 180 | 75 | |
XM_012962599.2 | XP_012818053.1 | 2 - 3 | 308 | 75 | ||
XM_018093394.1 | XP_017948883.1 | 2 - 3 | 260 | 75 | ||
GenBank mRNA | BC157801.1 | - | 2 - 3 | 258 | 75 | |
CR760329.2 | - | 2 - 3 | 180 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |