Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016725.2 | NP_001016725.1 | 2 - 3 | 617 | 80 | |
GenBank mRNA | BC121521.1 | - | 2 - 3 | 590 | 80 | |
CR760058.2 | - | 2 - 3 | 617 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |