Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016816.2 | NP_001016816.1 | 7 - 8 | 892 | 103 | |
GenBank mRNA | CR855555.2 | - | 7 - 8 | 892 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |