Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spryd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016934.2 | NP_001016934.1 | 1 - 2 | 101 | 83 | |
XM_012957010.2 | XP_012812464.1 | 2 - 3 | 757 | 83 | ||
GenBank mRNA | CR762113.2 | - | 1 - 2 | 101 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |