Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: leprot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017102.2 | NP_001017102.1 | 3 - 4 | 344 | 75 | |
GenBank mRNA | CR761280.2 | - | 3 - 4 | 344 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |