Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017167.2 | NP_001017167.1 | 4 - 5 | 349 | 84 | |
GenBank mRNA | CR760786.2 | - | 4 - 5 | 349 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |