Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017261.3 | NP_001017261.1 | 1 - 2 | 121 | 71 | |
GenBank mRNA | BC135935.1 | - | 1 - 2 | 127 | 71 | |
CR760012.2 | - | 1 - 2 | 124 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |