Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017279.2 | NP_001017279.1 | 517 | 61 | ||
GenBank mRNA | CR855741.2 | - | 517 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Xenopus tropicalis |