Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203604.2 | NP_988935.2 | 6 - 7 | 720 | 76 | |
XM_012970235.2 | XP_012825689.1 | 7 - 8 | 938 | 76 | ||
GenBank mRNA | BC061409.1 | - | 6 - 7 | 76 | ||
CR848649.2 | - | 6 - 7 | 720 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |