Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203964.3 | NP_989295.2 | 2 - 3 | 319 | 83 | |
GenBank mRNA | BC064234.1 | - | 2 - 3 | 226 | 83 | |
CN080459.1 | - | 2 - 3 | 256 | 83 | ||
CR926434.1 | - | 2 - 3 | 300 | 83 | ||
CX876971.1 | - | 2 - 3 | 319 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |