Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204041.2 | NP_989372.1 | 2 - 3 | 330 | 66 | |
GenBank mRNA | BC064191.1 | - | 2 - 3 | 327 | 66 | |
CR855594.2 | - | 2 - 3 | 330 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |