Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011031.2 | NP_001011031.1 | 3 - 4 | 120 | 55 | |
GenBank mRNA | BC082823.1 | - | 3 - 4 | 134 | 55 | |
CR760583.2 | - | 3 - 4 | 95 | 55 | ||
CR761935.2 | - | 3 - 4 | 120 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |