Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016068.3 | NP_001016068.2 | 4 - 5 | 537 | 84 | |
GenBank mRNA | BC091075.1 | - | 4 - 5 | 510 | 84 | |
BC118749.1 | - | 4 - 5 | 537 | 84 | ||
CR762257.2 | - | 4 - 5 | 600 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |