Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b4galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016211.3 | NP_001016211.1 | 4 - 5 | 658 | 68 | |
XM_012964682.1 | XP_012820136.1 | 5 - 6 | 625 | 68 | ||
XM_018094645.1 | XP_017950134.1 | 4 - 5 | 437 | 68 | ||
XM_018094646.1 | XP_017950135.1 | 4 - 5 | 425 | 68 | ||
GenBank mRNA | BC122889.1 | - | 4 - 5 | 658 | 68 | |
CR926330.2 | - | 5 - 6 | 598 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |