Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045633.1 | NP_001039098.1 | 2 - 3 | 199 | 82 | |
GenBank mRNA | BC159285.1 | - | 2 - 3 | 548 | 82 | |
CR942380.2 | - | 2 - 3 | 199 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |