Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: admp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045692.1 | NP_001039157.1 | 4 - 5 | 781 | 79 | |
GenBank mRNA | CR942593.2 | - | 4 - 5 | 781 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |