Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mogat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045737.1 | NP_001039202.1 | 6 - 7 | 942 | 86 | |
XM_012970512.2 | XP_012825966.1 | 6 - 7 | 1005 | 86 | ||
GenBank mRNA | CR942398.2 | - | 6 - 7 | 942 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |