Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mcm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045766.1 | NP_001039231.1 | 3 - 4 | 469 | 78 | |
GenBank mRNA | BC167938.1 | - | 3 - 4 | 460 | 78 | |
CR942676.2 | - | 3 - 4 | 469 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |