Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078750.1 | NP_001072218.1 | 6 - 7 | 711 | 78 | |
XM_018091333.1 | XP_017946822.1 | 6 - 7 | 792 | 78 | ||
GenBank mRNA | BC118796.1 | - | 6 - 7 | 711 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |