Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078814.1 | NP_001072282.1 | 4 - 5 | 740 | 64 | |
XM_018096301.1 | XP_017951790.1 | 4 - 5 | 601 | 64 | ||
GenBank mRNA | BC121300.1 | - | 4 - 5 | 740 | 64 | |
BC135462.1 | - | 4 - 5 | 680 | 64 | ||
CR762328.2 | - | 4 - 5 | 708 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |