Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078817.1 | NP_001072285.1 | 15 - 16 | 2949 | 67 | |
XM_012952548.2 | XP_012808002.1 | 15 - 16 | 3503 | 67 | ||
GenBank mRNA | BC121308.1 | - | 15 - 16 | 2949 | 67 | |
CR926259.1 | - | 3 - 4 | 316 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |