Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078823.1 | NP_001072291.1 | 1 - 2 | 1595 | 99 | |
GenBank mRNA | BC121322.1 | - | 1 - 2 | 1595 | 99 | |
CT025291.2 | - | 1 - 2 | 1597 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |