Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078873.1 | NP_001072341.1 | 624 | 73 | ||
XM_012965665.2 | XP_012821119.1 | 4 - 5 | 801 | 73 | ||
GenBank mRNA | BC121423.1 | - | 624 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |