Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078891.1 | NP_001072359.1 | 2 - 3 | 611 | 105 | |
GenBank mRNA | BC121458.1 | - | 2 - 3 | 611 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |