Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078925.1 | NP_001072393.1 | 7 - 8 | 693 | 83 | |
GenBank mRNA | BC121590.1 | - | 7 - 8 | 693 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |