Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078926.1 | NP_001072394.1 | 4 - 5 | 434 | 66 | |
GenBank mRNA | BC121591.1 | - | 4 - 5 | 434 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |