Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078931.2 | NP_001072399.2 | 2 - 3 | 738 | 93 | |
XM_012959240.2 | XP_012814694.1 | 2 - 3 | 1219 | 93 | ||
GenBank mRNA | BC121602.1 | - | 2 - 3 | 738 | 93 | |
BC160471.1 | - | 2 - 3 | 776 | 93 | ||
CU075314.1 | - | 2 - 3 | 729 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |