Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pyy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078959.1 | NP_001072427.1 | 3 - 4 | 299 | 101 | |
XM_012952555.2 | XP_012808009.1 | 4 - 5 | 1356 | 101 | ||
XM_012952556.2 | XP_012808010.1 | 3 - 4 | 379 | 101 | ||
XM_018097778.1 | XP_017953267.1 | 5 - 6 | 548 | 101 | ||
GenBank mRNA | BC121668.1 | - | 3 - 4 | 299 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |