Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078981.1 | NP_001072449.1 | 4 - 5 | 705 | 92 | |
XM_012952558.2 | XP_012808012.1 | 4 - 5 | 814 | 92 | ||
XM_012952559.2 | XP_012808013.1 | 4 - 5 | 809 | 92 | ||
GenBank mRNA | BC121706.1 | - | 4 - 5 | 705 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |