Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079206.1 | NP_001072674.1 | 12 - 13 | 1524 | 69 | |
XM_012957298.2 | XP_012812752.1 | 11 - 12 | 1751 | 69 | ||
GenBank mRNA | BC124001.1 | - | 12 - 13 | 1524 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |