Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079218.1 | NP_001072686.1 | 6 - 7 | 570 | 84 | |
GenBank mRNA | BC124019.1 | - | 6 - 7 | 570 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |