Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paqr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079240.1 | NP_001072708.1 | 6 - 7 | 646 | 87 | |
XM_012959271.2 | XP_012814725.1 | 7 - 8 | 925 | 87 | ||
XM_012959273.2 | XP_012814727.1 | 6 - 7 | 716 | 87 | ||
XM_012959274.2 | XP_012814728.1 | 6 - 7 | 741 | 87 | ||
XM_012959275.2 | XP_012814729.1 | 6 - 7 | 737 | 87 | ||
XM_018092178.1 | XP_017947667.1 | 7 - 8 | 921 | 87 | ||
GenBank mRNA | BC124054.1 | - | 6 - 7 | 646 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |