Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079262.2 | NP_001072730.2 | 4 - 5 | 1257 | 99 | |
GenBank mRNA | BC125705.1 | - | 4 - 5 | 1257 | 99 | |
CT025323.2 | - | 4 - 5 | 1164 | 99 | ||
CX740928.1 | - | 433 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |